Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System.
Gibco™

Neurobasal™-A Medium, minus phenol red

Catalog number:  12349015

 Related applications: Mammalian Cell Culture | Primary Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Neurobasal Medium minus phenol red is a basal medium designed for long-term maintenance and maturation of pure post-natal and adult brain neurons without the need for an astrocyte feeder layer when used with Gibco B-27 supplements. Use this modified formulation of Neurobasal Medium when the presence of phenol red will interfere with your results.

Neurobasal Medium is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of Neurobasal media and B-27 supplements.

For established protocols using Neurobasal media, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Tests Performed: Endotoxin, Osmolality, Performance, Sterility, pH
Cell Type: Neural Cells
Classification: Serum-Free
Product Size: 500 mL
Shipping Condition: Ambient

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products