Platinum™ SuperFi™ Green DNA Polymerase

Numéro de catalogue:  12357010

Invitrogen™

Error lors du chargement de votre contenu!

  Numéro de catalogue
Choisir votre mod�le de prix
Taille unitaire
Prix ({{currency}}) Disponibilité Qté
{{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} 
{{product.availability.message}}
Ajouts

Votre stock sur site

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} En stock)
›› {{supplyCenter.scName}}(En rupture de stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Haut de la page

Description

Invitrogen™ Platinum™ SuperFi™ Green DNA Polymerase provides Platinum™ SuperFi™ DNA Polymerase and a 5X SuperFi™ Green Buffer. The SuperFi Green Buffer contains a density reagent and two tracking dyes for convenient direct gel loading of PCR products.

Benefits of Platinum SuperFi DNA Polymerase include:
• Exceptional >100X Taq fidelity
• High specificity and increased yields with Platinum hot-start technology
• Robust amplification of difficult-to-amplify targets including those of sub-optimal purity or with ˃65% GC content
• Convenient workflow with room temperature reaction setup and 24 h bench top stability of pre-assembled reactions

Platinum SuperFi DNA Polymerase is a proofreading DNA polymerase that combines superior fidelity with trusted Platinum™ hot-start technology for the highest success in PCR. Featuring >100X Taq fidelity, Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Applications of Platinum SuperFi DNA Polymerase

• High-fidelity PCR
• Cloning and sub-cloning
• Site-directed mutagenesis
• Amplification of GC-rich templates
• Template generation for sequencing
• High-throughput PCR
• Amplification of samples with suboptimal purity
• Long PCR
• Fast PCR

Using Platinum SuperFi Green DNA Polymerase
Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases (such as Taq DNA polymerases). For optimal results, use the Tm calculator on our website: www.thermofisher.com/tmcalculator.

Platinum SuperFi Green DNA Polymerase is supplied with a separate vial of SuperFi™ GC Enhancer designed for difficult and GC-rich templates (˃65% GC). The SuperFi Green buffer that is also included contains a density reagent and two electrophoresis tracking dyes allowing direct loading of PCR products on a gel. The dyes in the green buffer do not interfere with PCR performance and are compatible with downstream applications including DNA sequencing, ligation, and restriction digestion.

Platinum hot-start technology efficiently inhibits DNA polymerase activity at ambient temperatures, allowing convenient reaction setup at room temperature. In addition, pre-assembled PCR reactions may be stored at room temperature for up to 24h prior to the PCR.

More Platinum SuperFi DNA Polymerase products ›

For Research Use Only. Not for use in diagnostic procedures.

Spécifications

Polymerase: Platinum SuperFi DNA Polymerase
Hot Start: Built-In Hot Start
Fidelity (vs. Taq): 100 X
GC-Rich PCR Performance: High
Reaction Format: Separate Components
Product Overhang: Blunt
Reaction Speed: Fast
Product Line: Platinum™, SuperFi™
Product Size: 100 units
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contenu et stockage

• 50 µL Platinum SuperFi DNA Polymerase (2U/µL)
• 2 x 1.25 mL SuperFi Green Buffer (5X)
• 1.25 mL SuperFi GC Enhancer (5X)

Store at -5 to -30°C.

Documents

Manuels et protocoles

Produits recommandés