Invitrogen™

DETACHaBEAD™ CD19 Kit

Catalog number:  12506D

 Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

B cells isolated with Dynabeads® CD19 and released with DETACHaBEAD® CD19 are pure, viable, phenotypically unaltered and unactivated. Cells are suitable for applications such as further sorting, culturing studies, and flow cytometry.

This Product Contains: DETACHaBEAD® CD19, polyclonal anti-Fab to release cells from Dynabeads® CD19 (Pan B). For use with cells isolated using Dynabeads® CD19
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: B Cells
Isolation Method: Positive Isolation
Output Viability: >95%
Product Line: DETACHaBEAD®, Dynabeads®
Product Size: 5 mL
Reactivity: Human
Sample Type (Specific): Blood, PBMC
Starting Material (Cell #): 1x10^7 PBMCs per isolation
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products