PCR Cloning System with Gateway® Technology with pDONR™/Zeo & OmniMAX™2 Competent Cells

Catalog number: 12535037

Invitrogen™  Related applications: Gateway Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gateway® Technology enables rapid cloning of one or more genes into virtually any protein expression system. Once you have an entry clone, you can recombine your gene of interest into a variety of expression vectors adapted for use in Gateway® Technology. The PCR Cloning System with Gateway® Technology is designed to create an entry clone following PCR with attB-modified custom primers (Figure 1). The PCR product is directionally cloned with efficiencies typically >99%. The PCR Cloning System with Gateway® Technology offers:

• Rapid, efficient, directional cloning of PCR products
• Versatility to use any amplification enzyme
• Cloning without the need for restriction endonucleases or ligase
• Selection of entry clones with kanamycin or Zeocin™

In addition, the pDONR™221 and pDONR™/Zeo vectors have a pUC origin for high plasmid yields and universal M13 sequencing sites for ease of use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Vector Type: pDONR
Cloning Method: Gateway®
Product Line: Gateway®, OmniMAX™, pDONR™
Product Size: 20 reactions

Contents & storage

The PCR Cloning Kit with Gateway® Technology includes Gateway® BP Clonase® II enzyme mix and reaction buffer, Gateway® pDONR™221 or pDONR™/Zeo vector, M13 sequencing primers, PEG/Mg2+ solution, proteinase K, pEXP7-tet positive control, and One Shot® OmniMAX™2 Competent Cells. Store competent cells and enzyme mix at -80°C. Store all other components at -20°C.

Documents

Manuals & protocols

Recommended products