Lipofectamine® 2000 CD Transfection Reagent

Catalog number: 12566014

Invitrogen™  Related applications: Antibody & Cell Culture Production | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Lipofectamine® 2000 CD (Chemically Defined) Transfection Reagent is a proprietary animal origin-free formulation for transfecting nucleic acids into eukaryotic cells. Using Lipofectamine® 2000 CD reagent for transfection provides the following advantages:

• Highest transfection efficiency in many cell types and formats.
• The animal origin-free formulation ensures that mammalian cell culture and bioproduction processes are free of animal-derived materials.
• DNA-Lipofectamine® 2000 CD complexes can be added directly to cells in culture medium.
• It is not necessary to remove complexes or change/add medium after transfection, but complexes may be removed after 4–6 hours.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines
Sample Type (Specific): Plasmid DNA
Classification: Animal Origin-Free, Chemically Defined
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
Product Size: 1 mL
Shipping Condition: Wet Ice
High Throughput Compatibility: Not High Throughput-Compatible (Manual)

Contents & storage

Each reagent should be stored at 2–8°C upon receipt. Reagents are guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products