GeneBLAzer™ In Vivo Detection Kit
Catalog number: 12578134
Invitrogen™  Related applications: Cellular Imaging | Protein Assays and Analysis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The GeneBLAzer™ In Vivo Detection Kit measures β-lactamase reporter activity in live mammalian cells, using the β-lactamase substrate CCF2-AM. The GeneBLAzer™ Detection System provides the following advantages:
• Suitable for use as a sensitive reporter of gene expression in living mammalian cells using fluorescence microscopy.
• Provides a ratiometric readout to minimize differences due to variability in cell number, substrate concentration, fluorescence intensity, and emission sensitivity
• Uses a non-toxic substrate, allowing continued cell culturing after quantitative analysis.
• Compatible with a wide variety of applications, including microplate-based transcriptional assays and flow cytometry.
• Provides a flexible and simple assay development platform for gene expression in mammalian cells.
For Research Use Only. Not for use in diagnostic procedures.
Figures
Specifications
Detection Method: Fluorescent
Product Line: GeneBLAzer®
Product Size: 375 reactions
Reporter: BLA (Beta-Lactamase)
Substrate: CCF2-AM
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Lactamase Substrate
Contents & storage
In vivo detection kits contain 50 µg of CCF2-AM substrate (enough for 375 96-well reactions). Substrate should be stored at -20°C, dessicated, and protected from light. All reagents are guaranteed stable 6 months when properly stored.
Documents
Manuals & protocols
Recommended products