Interested in a bulk or custom version of this product? Request pricing.

FABP5 Recombinant Human Protein

Catalog number: 12581HNAE5

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Fatty Acid Binding Protein 5 (psoriasis-associated) (FABP5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human FABP5 (Q01469) (Met 1-Glu 135).

Activity: This recombinant protein has not been tested.

Formulation: 50 mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Expression: E. coli
Form: Lyophilized
Gene Aliases: E-FABP, EFABP, PA-FABP, PAFABP
Gene ID: 2171
Gene Symbol: FABP5
Molecular Weight: 15.2 kDa
Product Line: Sino Biological®
Product Size: 5 µg
Protein Family: Other Proteins
Protein Form: Full Length, Recombinant
Protein Name: Fatty Acid Binding Protein 5 (psoriasis-associated)
Protein Tag or Fusion: Untagged
Purification: PAGE
Purity: ≥ 90% by SDS-PAGE
Research Category: Cardiovascular Research
Species: Human
Tag Position: None
Validated Application: Assay Standard

Contents & storage

Includes 1 vial of lyophilized protein. Store at -20 °C.

Documents

Manuals & protocols