Gibco™

Adenovirus Expression Medium (AEM), for PER.C6™ cells This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  12582011

 Related applications: Mammalian Cell Culture | Protein Expression | Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Adenovirus Expression Medium (AEM) is recommended for the growth of PER.C6® cells in suspension and for the production of viruses and viral vectors in these cells. It is an animal-origin free cell culture medium without components directly derived from human or animal sources. Each lot of AEM is performance tested for cell growth with PER.C6® cells and AEM has been qualified to produce adenovirus and influenza viruses. It also supports HEK 293 cell growth. Glutamine or GlutaMAX™-I supplementation is required.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Line: 293 (E1), 293 (HEK), PER.C6
Classification: Animal Origin-Free, Serum-Free
Culture Type: Suspension Cell Culture
Form: Liquid
Glutamine: No Glutamine
Product Line: Gibco®
Product Size: 1,000 mL
Serum Level: Serum-Free
Species: Human
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store at 2°C to 8°C, in the dark.

Documents

Manuals & protocols

Recommended products