Gibco™

Insulin, human recombinant, zinc solution

Catalog number:  12585014

 Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

4 mg/ml

Potency: (dry basis) >= 27 USP units/mg.
Suggested range for supplementation: 2 to 20 µg/ml.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Size: 5 mL
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products