Gibco™

CTS™ KnockOut™ SR XenoFree Medium This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  12618012

 Related applications: Clinical & Translational Research | Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

GIBCO® CTS™ KnockOut™ SR XenoFree Medium, when formulated as a complete medium, enables the growth and expansion of human embryonic stem cells (hESC) and human induced pluripotent stem cells (hiPSC) in a cell culture medium containing only human-derived or human recombinant proteins, to facilitate the transition of hESC and hiPSC research from the bench to the clinic.

The GICTS™ product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.

• Defined, xenofree medium for hESC⁄iPSC stem cell culture.
• First-to-market xenofree hESC⁄iPSC product intended for Ex Vivo tissue and cell culture processing applications.

CTS™ KnockOut™ SR XenoFree Medium does not contain bovine or other non-human, animal-derived components. Besides pluripotent stem cell culture expansion and maintenance, CTS™ KnockOut™ SR XenoFree Medium can be used for hESC⁄hiPSC cryopreservation, derivation, and differentiation studies.

GIBCO® CTS™ products are of high quality, and are supplied with harmonized documentation such as Certificates of Analysis, Certificates of Origin, and access to authorization letters for our Drug Master Files, as appropriate.
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.

Specifications

Manufacturing Quality: cGMP for medical devices, 21 CFR Part 820 and ISO 13485
Classification: Serum-Free, Xeno-Free
Cell Type: Stem Cells (Embryonic), Stem Cells (iPS - Induced Pluripotent Stem)
Form: Frozen
Glutamine: No Glutamine
Product Line: CTS™, KnockOut™
Product Size: 100 mL
Purity or Quality Grade: ES Cell-Qualified
Species: Human
Sterility: Sterile
Green Features: Sustainable packaging
Shipping Condition: Dry Ice

Contents & storage

-5 to -20°C

Documents

Manuals & protocols