Fetal Bovine Serum, mesenchymal stem cell-qualified, One Shot™ format, USDA-approved regions

Catalog number: 12662002

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sera Category: Specialty
Fetal Bovine Serum, MSC-Qualified One Shot™ is specially tested for the ability to support the expansion and clonal enumeration (Colony Forming Unit Fibroblast Assay) of mesenchymal stem cells. It’s the GIBCO® FBS quality and performance you trust offered in single-use 50-ml FBS aliquots which provide greater convenience, help save time, are easy to use, enable reproducibility, and reduce the risk of contamination.

Specifications

Age: Fetal
Format: Bottle(s)
Species: Cattle⁄Bovine
Donor Herd: No
Culture Type: Adherent Cell Culture
Product Size: 50 mL
Endotoxin Level: ≤ 50 EU/ml
Serum Treatment: Standard (Sterile-Filtered)
Tests Performed: Blue Tongue Virus
Volume (Metric): 50 ml
Country of Origin: USDA-Approved
Purity or Quality Grade: MS Cell-Qualified
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols