Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions

Catalog number: 12662011

Gibco™  Related applications: Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sera Category: Specialty
Specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.

Origin: Countries meeting USDA importation requirements.

Endotoxin level: <=50 EU/ml (levels routinely <=10 EU/ml).

Hemoglobin level: <=25 mg/dl.

Storage:
Store at -5°C to -20°C.

Intended use(s):
in vitro diagnostic (IVD).

Specifications

Age: Fetal
Format: Bottle(s)
Species: Cattle⁄Bovine
Product Size: 100 mL
Serum Treatment: Standard (Sterile-Filtered)
Country of Origin: USDA-Approved
Purity or Quality Grade: MS Cell-Qualified
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products