LHC Basal Medium (1X)

Catalog number: 12677027

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.

LHC Basal Medium (1X), liquid

Applications:
Formulated for the growth of bronchial epithelial cells with further supplements.

Recommended storage conditions:
2° to 8°C.

Shelf life:
12 months

Intended use(s)
Research use only



LHC-8 Medium and LHC-9 Medium


Applications:


Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.


Performance and quality testing:

Performance tested in a growth assay using BEAS-2B cells.

Recommended storage conditions:
-5°C to -20°C.

Intended use(s)
Research use only

Reference(s):
Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Form: Liquid
Species: Cattle⁄Bovine, Human
Cell Type: Epithelial Cells (Bronchial)
Glutamine: L-Glutamine
Serum Level: Serum-Free
Product Size: 1,000 mL
Classification: Serum-Free
Phenol Red Indicator: Phenol Red
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products