Interested in a bulk or custom version of this product? Request pricing.

PDE2A Recombinant Human Protein, His Tag

Catalog number: 13031H08B250

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Phosphodiesterase 2A,cGMP-Stimulated (PDE2A), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Glu 215-His 900) of human PDE2A (O00408-1) fused to a polyhistidine tag at the C-terminus.

Activity: The activity of this protein has not been tested.

Formulation: 20 mM Tris, 500 mM NaCl, 10% Glycerol, pH 7.4.

Reconstitution: No reconstitution needed. Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by Sino Biological Inc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Protein Family: Other Proteins
Research Category: Cardiovascular Research, Signal Transduction
Endotoxin Level: < 1 EU/µg
Expression: Insect
Form: Frozen
Gene Aliases: PDE2A1, PED2A4
Gene ID: 5138
Gene Symbol: PDE2A
Molecular Weight: 80 kDa
Product Line: Sino Biological®
Product Size: 5 x 50 µg
Protein Form: Recombinant, Truncated
Protein Name: phosphodiesterase 2A, cGMP-stimulated
Protein Tag or Fusion: His Tag
Purification: PAGE
Purity: ≥ 85% by SDS-PAGE
Species: Human
Validated Application: Assay Standard

Contents & storage

Includes 5 vials of protein in suspension buffer. Store at -80 °C.

Documents

Manuals & protocols

Recommended products