Nalgene™ Plastic Labware Value Pack
Thermo Scientific™  Related applications: Bottles and Accessories
Choose from an assortment of popular, everyday labware with the Thermo Scientific™ Nalgene™ Plastic Labware Value Pack, perfect as a new lab starter or refresher pack. Nalgene Labware sets the standard for quality and superior performance for your valuable research with the highest quality materials for dependable performance.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Introducing shared lists
Back to top
  • Receive 20 items with one catalog number
  • A variety of everyday lab items at one low price
  • Won't shatter or break
Six straight-sided jars (500mL/PPCO), four unitary wash bottles (500mL/LDPE), four graduated cylinders (2-100mL, 500mL, 1L/PPCO), five Griffin low form beakers (50, 100, 250, 600, 1000mL/PPCO), Thermo Scientific™ Nalgene™ Unwire™ blue half-rack for 30mm tubes.
Remove jar closure and autoclave separately. LDPE unitary wash bottles are not autoclavable. All other items in Value Pack are autoclavable.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Manuals & protocols