See the new Lipofectamine® RNAiMAX Transfection Reagent protocol—now optimized for efficiency, viability and reproducibility
New Transfection Support Center—find technical tips and troubleshooting recommendations from our scientists.
New RNAi Support Center—Find technical tips and troubleshooting recommendations for your RNAI experiments from our scientists.

Lipofectamine® RNAiMAX Transfection Reagent This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 13778075

Invitrogen™  Related applications: RNAi | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Lipofectamine® RNAiMAX Transfection Reagent provides the highest transfection efficiencies on the widest variety of cell types for siRNA-mediated gene knockdown experiments. Lipofectamine® RNAiMAX is a proprietary RNAi-specific cationic lipid formulation designed specifically for the delivery of siRNA and miRNA into all cell types.

With Lipofectamine® RNAiMAX Transfection Reagent you will get:

• Superior transfection efficiency requiring lower RNAi concentrations, leading to more effective gene knockdown with minimal nonspecific effects
• Easy optimization due to minimal cytotoxicity across a 10-fold concentration range of transfection reagent
• Superior transfection efficiencies for miRNA antagonists and mimics
• Compatibility with a broad range of cell types, providing the most versatile approach to all of your gene silencing experiments
• A simple and rapid protocol for consistent and reproducible results

High knockdown in a wide range of cells
Lipofectamine® RNAiMAX Transfection Reagent transfects a wide range of cell types (see figure). For gene silencing, Lipofectamine® RNAiMAX Transfection Reagent's high-efficiency transfections lead to the high levels of gene knockdown needed to achieve convincing results.

Simple, high-throughput–ready transfections
Simply mix Lipofectamine® RNAiMAX Transfection Reagent with siRNA, add to your cells, incubate, and measure gene knockdown. The simplicity and speed combined with high transfection efficiency make Lipofectamine® RNAiMAX Transfection Reagent ideal for high-throughput siRNA transfections. Transfection conditions can be easily established for automated or robotic systems used in such applications.

Find an optimized Lipofectamine® RNAiMAX transfection protocol for your cell line.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines, Hard-to-Transfect Cells, Primary Cells, Stem Cells
Sample Type (Specific): Synthetic siRNA
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
High Throughput Compatibility: High Throughput-Compatible
Product Line: Lipofectamine®
Product Size: 0.75 mL
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice
Green Features: Sustainable packaging

Contents & storage

Contains one vial (0.75 ml) Lipofectamine® RNAiMAX Transfection Reagent. Store at 2–8°C. Do not freeze.

Documents

Manuals & protocols

Recommended products