Invitrogen™

Dynabeads™ M-450 Tosylactivated

Catalog number:  14013

 Related applications: Cell Isolation & Expansion | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dynabeads® M-450 Tosylactivated are 4.5 µm superparamagnetic beads containing surface tosyl groups. The beads covalently bind primary amino and sulfhydryl groups in antibodies to position them in the optimal orientation for cell-surface protein binding. Low background and covalent binding of the antibodies make these beads an excellent choice for cell separation applications when a downstream release from the beads is not required.

• Quickly deplete cells or positively isolate target cells for downstream molecular analysis
• Excellent for cell isolation procedures using your own unique antibody
• Several antibodies can be coupled on the same beads for non-cell-based applications, such as cell activation

Coupling procedure
Covalent coupling is performed overnight by incubating the antibody of choice with the Dynabeads® M-450 Tosylactivated beads. Optimal coupling occurs at high pH (8.5–9.5) and at 37°C. For pH-labile antibodies, coupling can be performed in an alternative buffer at pH 7.4.

Not recommended for immunoprecipitation (IP)
These 4.5 µm Tosylactivated beads are not recommended for IP as they have a lower surface capacity per unit mass when compared to 1 µm and 2.8 µm Tosylactivated and Epoxy Dynabeads®.

Learn more about Dynabeads® products
• Find Dynabeads® products for a whole range of applications.
• Find magnets for Dynabeads® separations.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~2x10^9 cells total
Cell Type: All cells from all species
Isolation Method: Depletion, positive isolation for molecular applications
Output Viability: >95%
Product Line: DYNAL®, Dynabeads®
Product Size: 5 mL
Reactivity: All species
Sample Type (Specific): Blood, PBMC, Tissue Digests
Starting Material (Cell #): 1x10^7 cells per isolation
Target Species: All species
Shipping Condition: Room Temperature

Contents & storage

Contains: 5 mL beads at a concentration of 4 × 10^8 beads/ml in distilled water.
Storage: 2°C to 8°C.

Documents

Manuals & protocols

Recommended products