DPBS, no calcium, no magnesium

Catalog number: 14190342

Gibco™  Related applications: Antibody & Cell Culture Production | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

We offer a variety of Gibco® DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

Without
• Calcium
• Magnesium
• Phenol Red

The complete formulation is available.

cGMP Manufacturing and Quality System
Gibco® DPBS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco® DPBS product made in our Grand Island facility (14190-144). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

Specifications

Form: Liquid
Inorganic Salts: No Calcium, No Magnesium
Concentrated: 1 X
Phenol Red Indicator: No Phenol Red
pH Range: 7.0 - 7.3
Osmolality: 270 - 300 mOsm/kg
Product Size: 5 L
Classification: Animal Origin-Free
Sodium Pyruvate Additive: No Sodium Pyruvate
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 15°C to 30°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture

Documents

Manuals & protocols

Recommended products