Dynabeads® M-270 Amine

Catalog number: 14308D

Invitrogen™  Related applications: Protein Assays and Analysis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

These are hydrophilic Dynabeads® (2.8 µm) with amine groups. The surface amino groups allow direct covalent binding of carbohydrates, glycoproteins and glycolipids through reductive amination of aldehydes.

• Rapid binding (less than 1 hour) at neutral to high pH and room temperature.
• No further surface activation required.
• Excellent reactivity with a range of cross-linking reagents, allowing for easy introduction of further alternative surface chemistries.

Applications:
Ideal for isolation of small components such as proteins and peptides. Allows C-terminal coupling of proteins. Well suited for isolation of proteins or other targets with affinity for specific carbohydrate moieties.

Concentration:
2 x 109 beads / ml

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Surface Functionality: Amine
Elution Volume - min: 5 µl
Form: Beads in Suspension
High Throughput Compatibility: High Throughput-Compatible
Product Line: Dynabeads®
Product Size: 10 mL
Sample Type (Specific): Any Sample Type
Shelf Life: 36 months from date of manufacture
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products