Penicillin-Streptomycin (10,000 U/mL)

Catalog number: 15140163

Gibco™  Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The antibiotics penicillin and streptomycin are used to prevent bacterial contamination of cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin was originally purified from the fungi Penicillium and acts by interfering directly with the turnover of the bacterial cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin was originally purified from Streptomyces griseus. It acts by binding to the 30S subunit of the bacterial ribosome, leading to inhibition of protein synthesis and death in susceptible bacteria. This solution contains 10,000 units/mL of penicillin and 10,000 µg/mL of streptomycin in a 10 mM citrate buffer (for pH stability).

We offer a wide range of antibiotics and antimycotics in both powder and liquid formats. See the complete list, or find products for:
Contamination control
Eukaryotic and bacterial selection

See recommended working concentrations for selection antibiotics.

Learn more about the use of antibiotics and antimycotics in cell culture and review guidelines for decontaminating cultures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Validated Application: Prevention of Cell Culture Contamination
Agent: Streptomycin, Penicillin
Form: Liquid
Reagent Type: Antibiotic
Concentrated: 100 X
Product Size: 20 x 100 mL

Contents & storage

Storage conditions: -5°C to -20°C
Shipping conditions: Dry ice
Shelf life: 12 months from date of manufacture

Documents

Manuals & protocols

Recommended products