This product is manufactured in our Grand Island, US facility and is intended for sale in North America, Latin America, and the Asia/Pacific region—higher freight charges will apply to orders shipped elsewhere. An equivalent product, manufactured in Paisley, Scotland, is recommended for our customers in Europe, Africa, and the Middle East. Click to view.
Gibco™

Distilled Water This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  15230204

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This distilled water is high-quality water for use as a solvent in the preparation of cell culture media and laboratory reagents. Available in many sizes to meet your needs, this distilled water features:

• Cell culture quality
• Proven performance
• Suitable for use in cell culture and molecular biology applications

Cell culture quality
This water is prepared by distillation and filtration (0.1 µm), with no added substances. We monitor the water quality on a routine basis. Conductivity, total organic carbon, endotoxin, and bioburden are tested daily; metals are tested monthly.

Proven performance
The performance and consistency expected from Gibco® liquid media products depends on the high-quality water we use in manufacturing. This water is also bottled as Gibco® Distilled Water.

Use in cell culture and molecular biology applications
This distilled water can be used for a wide variety of applications, including preparing powder or Advanced Granulation Technology™ (AGT™) media, making solutions, and diluting buffers.

Dual-site cGMP manufacturing and quality system
This water is produced at a cGMP-compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical product made in our Grand Island facility (15230-162). This facility is also registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Purity or Quality Grade: Cell Culture Grade, Endotoxin-Screened
Sample Treatment: Not DEPC-Treated
Format: Bottle(s)
Product Size: 10 x 500 mL
Purification: Membrane-Filtered
Sterility: Sterile
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 15°C to 30°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Documents

Manuals & protocols

Recommended products