Human Cot-1 DNA®

Catalog number: 15279011

Invitrogen™  Related applications: Cellular Imaging | Chromatin Biology | Genotyping & Genomic Profiling

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Human Cot-1 DNA® is commonly used to block nonspecific hybridization in microarray screening. It can also be used to suppress repetitive DNA sequences for the direct mapping of human DNA or mapping genomic clones to panels of somatic-cell hybrids for chromosome localization by Southern blotting. Human Cot-1 DNA® is effective as a library-screening probe for somatic-cell hybrid libraries and flow-sorted chromosome libraries made from somatic-cell hybrids.

About Cot-1 DNA®
Human Cot-1 DNA® is placental DNA that is predominantly 50 to 300 bp in size and enriched for repetitive DNA sequences such as the Alu and Kpn family members. The amount supplied per package is sufficient for 5–10 Southern or 500 in situ hybridizations.

Performance and quality testing
Purity and DNA size are verified by agarose gel electrophoresis. Concentration is verified spectrophotometrically by diluting Human Cot-1 DNA® 1:100 in 50 mM NaOH and using the conversion factor 0.033 µg/µL/A260. Other methods used to determine concentration may yield varying results.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Cot-1 DNA
Key Function: Block Non-Specific Hybridization
Product Line: Cot-1 DNA®
Product Size: 500 µg
Shipping Condition: Approved for shipment on Wet or Dry Ice

Contents & storage

This DNA is supplied at 1 mg/ml in 10 mM Tris-HCl (pH 7.4), 1 mM EDTA.
Store at -20°C.

Documents

Manuals & protocols

Recommended products