Invitrogen™

UltraPure™ Formamide

Catalog number:  15515026

 Related applications: Cellular Imaging | Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ Formamide is commonly used for denaturing nucleic acids for sequencing gel electrophoresis, electron microscopy, and hybridization. A liquid at room temperature, UltraPure™ Formamide is vacuum distilled and packaged under dry nitrogen and can be used as supplied for many applications within three months of date of purchase. Because the breakdown products of formamide degrade nucleic acids, for sensitive applications formamide should be deionized by treatment with a mixed-bed ion-exchange resin immediately before use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Line: UltraPure™
Product Size: 500 g
Reagent Type: Formamide
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products