Ribonuclease Inhibitor, Cloned

Catalog number:  15518012

Invitrogen™  Related applications: Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ribonuclease Inhibitor, Cloned is a potent non-competitive inhibitor of neutral pancreatic ribonucleases.

Applications:
Cell-free translation systems (1). Reverse transcription of mRNA (2).

Source:
Recombinant, purified from E. coli.

Performance and Quality Testing:
No detectable contaminating activity is observed in DNA nicking, ribonuclease, and protease assays.

Unit Definition:
One unit inhibits 5 ng RNase A by 50% using cytidine 2´, 3´ cyclic monophosphate (cCMP) as the substrate (3).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

LoyaltyEligible: SKUEligibleForLoyalty
Product Size: 1,000 units
Shipping Condition: Dry Ice

Contents & storage

Ribonuclease Inhibitor, Cloned is supplied at 10,000 units/ml in 20 mM HEPES-KOH (pH 7.6), 50 mM KCl, 8 mM DTT, and 50% (v/v) glycerol. Store at -20°C.

Documents

Manuals & protocols

Recommended products