X-Gal

Catalog number: 15520034

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactoside) is a histochemical substrate for β-galactosidase. This substrate yields a blue precipitate upon hydrolysis, making it suitable for use in immunoblotting or immunocytochemical assays. Bacteria containing active β-galactosidase produce blue colonies when grown on plates containing X-Gal. X-Gal is supplied as white crystals and is soluble in dimethylformamide.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Permeability: Cell-Permeant
Detection Method: Colorimetric
Product Size: 100 mg
Substrate: X-Gal
Substrate Properties: Chemical Substrate
Substrate Type: Beta-Gal Substrate
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products