Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

UltraPure™ 1 M Tris-HCI Buffer, pH 7.5

Catalog number: 15567027
Invitrogen™

UltraPure™ 1 M Tris-HCI Buffer, pH 7.5

Catalog number: 15567027
Catalog Number
15567027
also known as 15567-027
Unit Size
1 L
Price (USD)
Price: 68.00
Online price: 65.65
Your price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
15567027
also known as 15567-027
1 L
Price: 68.00
Online price: 65.65
Your price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
UltraPure™ 1 M Tris-HCl Buffer, pH 7.5, is a premixed, pH-adjusted, sterile-filtered solution. Prepared as a 1 M concentrate, this buffer can be diluted to the desired concentration and used in molecular biology or general biochemistry applications.

Performance and quality testing
No DNase, RNase, or protease activity detected.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
1 M
Green Features
Sustainable packaging
pH
pH 7.5
Product Line
UltraPure™
Product Type
Tris-HCl Buffer
Quantity
1 L
Shipping Condition
Room Temperature
Form
Liquid

Contents & Storage

Store in refrigerator (2-8°C). UltraPure™ 1M Tris-HCl, pH 7.5 is guaranteed for 6 months from date of purchase unless otherwise indicated in product documentation.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text