Invitrogen™

UltraPure™ 0.5M EDTA, pH 8.0

Catalog number:  15575020

 Related applications: Chromatin Biology | DNA Extraction | Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ 0.5 M EDTA, pH 8.0 is an aqueous solution prepared by dissolving Na2EDTA•2H2O in distilled, deionized water and adjusting the pH to 8.0 with sodium hydroxide. This sterile-filtered solution is suitable for biochemistry or molecular biology applications requiring a chelator of divalent metal ions.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: EDTA
Form: Liquid
Product Line: UltraPure™
Product Size: 4 x 100 mL
Purity or Quality Grade: Molecular Biology Grade
pH: pH 8.0
Shipping Condition: Room Temperature

Contents & storage

Contents: 4 bottles (100 mL each) of UltraPure™ 0.5M EDTA, pH 8.0. Store at room temperature.

Documents

Manuals & protocols

Recommended products