UltraPure™ SSPE, 20X

Catalog number: 15591027

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ 20X SSPE is a prepared buffer concentrate formulated for use in nucleic acid hybridizations and blot transfer applications. Supplied in 1 L plastic bottles, or in a 4 L or 10 L stackable Cubitainer¤ Box, sterile-filtered UltraPure™ 20X SSPE contains 3.0 M NaCl, 0.2 M NaH2PO4, and 0.02 M EDTA at pH 7.4.

Performance and Quality Testing: No DNase or RNase activity detected.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentrated: 20 X
Form: Liquid
Product Line: UltraPure™
Product Size: 10 L
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products