UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v)

Catalog number:  15593049

Invitrogen™  Related applications: DNA Extraction | RNA Extraction

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ Phenol:Chloroform:Isoamyl Alcohol (25:24:1, v/v) is used in the purification of nucleic acids. This reagent consists of highly pure chloroform, isoamyl alcohol, and UltraPure™ Phenol saturated with Tris-HCl.

Using Phenol:Chloroform:Isoamyl Alcohol
When mixtures are extracted with Phenol:Chloroform:Isoamyl Alcohol, proteins are denatured and collected in the organic phase or at the interphase, while nucleic acids remain in the aqueous phase. UltraPure™ Phenol:Chloroform:lsoamyl Alcohol contains no preservatives. It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles.

Performance and quality testing
The appearance of the solution is evaluated at room temperature. No RNase or DNase activity is detected.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name: Phenols
Form: Liquid
Product Line: UltraPure™
Product Size: 400 mL
Purity or Quality Grade: Molecular Biology Grade
pH: pH 8.05
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products