UltraPure™ Herring Sperm DNA Solution

Catalog number: 15634017

Invitrogen™  Related applications: Chromatin Biology | Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ DNA solutions from Calf Thymus, Herring Sperm, or Salmon Sperm are ready-to-use, sheared DNA solutions that are used directly in the preparation of prehybridization and hybridization solutions. These prepared solutions are used to block the non-specific attachment of probe to the surface of a membrane. The DNA solutions are prepared from highly pure, phenol/chloroform-extracted DNA and DNase-free and RNase-free (DEPC-treated) distilled, deionized water and are sheared to an average size of <=2,000 bp. The concentration is adjusted to 10 mg/ml.

Performance and Quality Testing:

Concentration, size verification, viscosity, and ultraviolet absorbance (A260/A280) are analytically tested.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reagent Type: Herring Sperm DNA
Key Function: Block Non-Specific Hybridization
Product Line: UltraPure™
Product Size: 5 x 1 mL
Shipping Condition: Dry Ice

Contents & storage

Store at -20°C.

Documents

Manuals & protocols

Recommended products