This product has been Gibco FBS Fingerprinted. We developed a technology in partnership with Oritain, a global leader in verifying product origins. Based on geographical differences, we can create "fingerprints" of our FBS that connect them to their origin. Find out more ›

Fetal Bovine Serum, certified, US origin

Catalog number: 16000036

Gibco™  Related applications: Insect Cell Culture | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® Sera: Trusted Quality for Consistent Performance

Sera Category: Performance Plus
• Use for a broad range of cell types, especially sensitive cell lines
• Our most characterized sera with the lowest endotoxin and hemoglobin levels
• Endotoxin level: ≤ 5 EU/ml
• Hemoglobin level: ≤ 10 mg/dl
• Origin: United States

Gibco® Serum Delivers
• ISO 13485 certified, processed in FDA registered facilities
• Labeled for in-vitro diagnostics (compliant with highest level of USP sterility testing)
• Triple filtered at 0.1 µm
• Gibco® bottle is easier to use in the hood, reduces the risk of contamination and helps you perform cell culture more consistently
• Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications

Serum Treatment: Standard (Sterile-Filtered)
Purity or Quality Grade: Certified
Country of Origin: United States
Format: Bottle(s)
Age: Fetal
Species: Cattle⁄Bovine
Product Size: 100 mL
Endotoxin Level: ≤ 5 EU/ml
Hemoglobin Level: ≤ 10mg⁄dl
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products