This product has been Gibco FBS Fingerprinted. We developed a technology in partnership with Oritain, a global leader in verifying product origins. Based on geographical differences, we can create "fingerprints" of our FBS that connect them to their origin. Find out more ›

Fetal Bovine Serum, certified, One Shot™ format, US origin

Catalog number: 16000077

Gibco™  Related applications: Insect Cell Culture | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sera Category: Performance Plus
Origin: United States
Endotoxin level: ≤5 EU/ml
Hemoglobin level: ≤10 mg/dl

Specifications

Serum Treatment: Standard (Sterile-Filtered)
Purity or Quality Grade: Certified
Country of Origin: United States
Format: One Shot
Age: Fetal
Species: Cattle⁄Bovine
Product Size: 50 mL
Endotoxin Level: ≤ 5 EU/ml
Hemoglobin Level: ≤ 10mg⁄dl
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products