Invitrogen™

Dynabeads™ Epithelial Enrich

Catalog number:  16102

 Related applications: Cell Isolation & Expansion

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Enrich epithelial tumor cells directly from whole blood, bone marrow, or PBMC suspensions. Enriched tumor cells are lysed for mRNA isolation and RT-PCR amplification. The sensitivity of combined enrichment and RT-PCR detection can be as high as 1 tumor cell in 1 ml blood.

• Depletion of epithelial tumor cells from whole blood/bone marrow or PBMC.
• Positive isolation of epithelial tumor cells for downstream molecular analysis.
• No columns required

Starting samples:
Whole blood, bone marrow, PBMC, buffy coat.

Number of cells processed:
200 ml whole blood or ~4 × 109 PBMC.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: Processes ~4x10^9 cells total
Cell Type: Epithelial Cells, Tumor Cells
Isolation Method: Depletion, positive isolation for molecular applications
Output Viability: >95%
Product Line: DYNAL®, Dynabeads®
Product Size: 5 mL
Reactivity: Human
Sample Type (Specific): Blood, PBMC
Starting Material (Cell #): 2x10^7 PBMCs per isolation
Target Species: Human
Shipping Condition: Room Temperature

Contents & storage

This Product Contains: 5 ml Dynabeads® coated with the monoclonal antibody BerEP4 against the human
epithelial antigen, EpCAM.
Store at 2-8°C

Documents

Manuals & protocols

Recommended products