Gibco™

Fetal Bovine Serum, qualified, heat inactivated, US origin This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  16140089

 Related applications: Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Sera Category: Performance
Origin: United States.
Endotoxin level: ≤10 EU/ml
Hemoglobin level: ≤25 mg/dl.

Specifications

Serum Treatment: Heat-Inactivated
Purity or Quality Grade: Qualified
Country of Origin: United States
Format: Bottle(s)
Endotoxin Level: ≤ 10 EU/ml
Hemoglobin Level: ≤ 25 mg⁄dl
Product Line: Gibco®
Product Size: 1,000 mL
Species: Cattle⁄Bovine
Green Features: Sustainable packaging
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products