Fetal Bovine Serum, embryonic stem cell-qualified, US origin This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 16141061

Gibco™  Related applications: Mammalian Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Sera Category: Specialty
Origin: United States.
Specially tested for the ability to sustain undifferentiated cellular morphology of embryonic stem cells.
Endotoxin level: <=50 EU/ml (levels routinely <= 10 EU/ml).
Hemoglobin level: <=25 mg/dl.

Specifications

Age: Fetal
Format: Bottle(s)
Species: Cattle⁄Bovine
Product Size: 100 mL
Serum Treatment: Standard (Sterile-Filtered)
Country of Origin: United States
Purity or Quality Grade: ES Cell-Qualified
Green Features: Fewer resources used, Less waste
Shipping Condition: Dry Ice

Contents & storage

Storage conditions: -5 to -20°C
Shipping conditions: Frozen

Documents

Manuals & protocols

Recommended products