New! Save technical documents to your personal library. Open a PDF, then look for the "Save to Library" icon.

FreeStyle™ MAX Reagent

Catalog number:  16447100

Gibco™  Related applications: Antibody & Cell Culture Production | Protein Expression | Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins. FreeStyle™ MAX Reagent allows the highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications, and is specifically formulated for use with:

• FreeStyle™ 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle™ 293 Expression Medium (Cat. No. 12338-018)
• FreeStyle™ CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle™ CHO Expression Medium (Cat. No. 12651-014)
• DG44 Cells (DHFR suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic® F-68 (Cat. No. A11000-01)

Product use
FreeStyle™ MAX Reagent is intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
For Research Use Only. Not for use in diagnostic procedures.


Cell Type: Established Cell Lines
Sample Type (Specific): Plasmid DNA
Classification: Animal Origin-Free
Transfection Technique: Lipid-Based Transfection
Serum Compatible: Yes
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: Freestyle™
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Each unit contains one vial of reagent. Store at +4°C. FreeStyle&trade MAX is guaranteed stable for 6 months when stored properly.


Manuals & protocols

Recommended products