UltraPure™ Low Melting Point Agarose This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 16520050

Invitrogen™  Related applications: Nucleic Acid Gel Electrophoresis & Blotting

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

UltraPure™ Low Melting Point Agarose is a polysaccharide used for size-based separation of nucleic acids in agarose gel electrophoresis applications. UltraPure™ Low Melting Point Agarose is ideal for resolving DNA and RNA fragments and for the recovery of nucleic acid fragments after electrophoresis, as it melts at 65.5°C (below the melting point of most nucleic acids). The solution remains fluid at 37°C and will set rapidly at temperatures below 25°C. Benefits of using UltraPure™ Low Melting Point Agarose:

• Ideal for analysis and recovery of DNA and RNA for routine applications
• Strong gel structure allows for better handling and less breakage
• Ideal for a variety of applications such as in-gel manipulation, tissue and cell culture, and viral plaque assays
• Can also be used in protein electrophoresis applications such as Ouchterlony (antigen-antibody interaction assay) and radial immunodiffusion (RID) (antigen quantitation assay)

Improved packaging
The new environmentally friendly packaging uses 75% less plastic than the original bottles (see figure). This means less energy and raw material used in manufacture, and less waste in landfills. Additionally, the easy-pour spout reduces the likelihood of spills and contamination.

Performance and quality testing
The performance of UltraPure™ Agarose is evaluated to satisfy specifications in appearance, moisture, gel strength, gelling temperature, melting temperature, sulfate content, electroendosmosis, and DNase/RNase assay.

Note: UltraPure™ Low Melting Point Agarose (Cat. No. 16520-100) is a replacement for the item previously sold under Cat. No. 15517-022.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Melting Point: Low Melting Point
Separation Range: 100 bp to >30 kb
Form: Powder
Gel Compatibility: Agarose Gels
Product Line: UltraPure™
Product Size: 50 g
Validated Application: Gel Electrophoresis, Plaque Assay
Green Features: Fewer resources used, Less waste
Shipping Condition: Room Temperature

Contents & storage

UltraPure™ Agarose products are packaged in new eco-friendly pouches that use less material than conventional plastic bottles. Store at 15° to 30°C.

Documents

Manuals & protocols

Recommended products