AmnioMAX™ C-100 Basal Medium

Catalog number: 17001082

Gibco™  Related applications: Cell Viability, Proliferation & Function | Clinical & Translational Research | Mammalian Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Gibco® AmnioMAX™ C-100 Basal Medium is part of a kit used to prepare a fully-supplemented Gibco® AmnioMAX™ C-100 Complete Medium developed for the short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures.

Gibco® AmnioMAX™ C-100 Complete Medium features:

• Easy-to-use format
• Quality and performance testing
• Unique, optimized formulation

Easy-to-use format
Gibco® AmnioMAX™ C-100 Complete Medium is a two-part kit for preparation of a complete medium that requires no additional supplementation. The complete medium can be stored at 2–8°C for up to ten days.

Quality and performance testing
Every lot of Gibco® AmnioMAX™ C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance. Pooled primary human amniotic fluid samples are cultured for six days in Gibco® AmnioMAX™ C-100 Complete Medium before measuring the total number of colonies and total number of mitotic colonies. In addition, each lot is tested for sterility, pH and osmolality.

Unique, optimized formulation
Gibco® AmnioMAX™ C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth. This optimized medium also has an enhanced buffering system that provides greater pH stability during culture manipulations.

Product Use
For in vitro diagnostic use.

cGMP Manufacturing and Quality System
Gibco® AmnioMAX™ C-100 Basal Medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”

Specifications

Form: Liquid
Species: Human
Cell Type: Amniocytes
Glutamine: L-Glutamine
Serum Level: Standard Serum
Product Size: 90 mL
Shipping Condition: Room Temperature

Contents & storage

Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 16 months from date of manufacture. Use complete medium within 7–10 days once opened.

Documents

Manuals & protocols

Recommended products