Increase cell survival by more than 50% with new Gibco™ B-27™ Plus Neuronal Culture System.
Taking your work to the clinic? Check out CTS N-2 »
Gibco™

N-2 Supplement (100X)

Catalog number:  17502048

 Related applications: Mammalian Cell Culture | Primary Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

N-2 Supplement is a chemically-defined, serum-free supplement based on Bottenstein’s N-1 formulation. It is recommended for growth and expression of neuroblastomas as well as post-mitotic neurons in primary cultures from both the peripheral nervous system and the central nervous system. N-2 Supplement is provided as a 100X concentrate and is intended for use with Neurobasal Medium supplemented with growth factors such as bFGF and EGF. It may also be used with DMEM.

For established protocols using N-2 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Neural Cells
Classification: Serum-Free
Product Size: 5 mL
Tests Performed: Endotoxin, Performance, Sterility
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -20°C) and protect from light.

Media formulations

Documents

Manuals & protocols

Help center

Cell Culture & Transfection Learning Center

Access cell culture and transfection educational resources for better experiment planning and execution.

Cell Culture & Transfection Support Centers

Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.

Recommended products