Gibco™

G-5 Supplement (100X)

Catalog number:  17503012

 Related applications: Mammalian Cell Culture | Primary Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

G-5 Supplement is a chemically defined, serum-free supplement based on Bottenstein’s G-5 formulation. It is recommended for growth and expression of glial cells (normal and tumor) of astrocytic phenotype (astrocytes). G-5 Supplement is provided as a 100X concentrate and is intended for use with Neurobasal Medium. It may also be used with DMEM, DMEM⁄F-12, or other basal media.

View publications using Gibco neurobiology products at our Gibco CellCite.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Neural Cells
Classification: Serum-Free
Product Size: 1 mL
Tests Performed: Endotoxin, Performance, Sterility
Shipping Condition: Dry Ice

Contents & storage

Store in freezer (-5 to -20°C) and protect from light.

Media formulations

Documents

Manuals & protocols

Recommended products