B-27™ Supplement (50X), serum free This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 17504001

Gibco™  Related applications: Mammalian Cell Culture | Primary Cell Culture | Stem Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

B-27® Supplement is an optimized serum-free supplement used to support the low or high density growth and short or long-term viability of hippocampal and other CNS neurons. B-27® Supplement is provided as a 50X liquid and is intended to be used with Neurobasal® medium or Neurobasal®-A medium for neuronal cell culture without the need for an astrocyte feeder layer.

B-27® Supplement is suitable for use in most neuronal cell applications. For specialized applications and alternative experimental needs, we provide other formulations of B-27® Supplement.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Neuronal Cells, Stem Cells (Neural)
Classification: Serum-Free
Product Line: B-27™
Product Size: 100 mL
Green Features: Fewer resources used, Less waste

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products