Liver Perfusion Medium (1X) This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number: 17701038

Gibco™  Related applications: Hepatocytes & Hepatic Models | Mammalian Cell Culture | Pharma & Biopharma | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Liver Perfusion Medium is a buffered, balanced salt solution formulated to cleanse the liver of blood, prevent clotting, and initiate loosening of cell-to-cell contact.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Hepatocytes
Classification: Serum-Free
Form: Liquid
Glutamine: No Glutamine
Phenol Red Indicator: No Phenol Red
Product Size: 500 mL
Serum Level: Serum-Free
Species: Human
Green Features: Fewer resources used, Less waste
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products