HepatoZYME-SFM This product, and/or its packaging, incorporates one or more environmental/green features. Click on this icon to learn more.

Catalog number:  17705021

Gibco™  Related applications: Hepatocytes & Hepatic Models | Mammalian Cell Culture | Pharma & Biopharma | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Hepatozyme-SFM is a serum-free medium for the long-term maintenance of hepatocyte phenotypic expression, including active and inducible forms of cytochromes P450 and active phase II enzymes.

Contains no phenol red.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Hepatocytes
Classification: Serum-Free
Form: Liquid
Glutamine: No Glutamine
Phenol Red Indicator: No Phenol Red
Product Size: 500 mL
Serum Level: Serum-Free
Species: Human
Green Features: Sustainable packaging
Shipping Condition: Room Temperature

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products