GFAP (Glial Fibrillary Acid Protein) Rabbit Polyclonal Antibody, Concentrate

Catalog number: 180063

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This polyclonal antibody stains GFAP. In general, normal and neoplastic astrocytic cells react positively for GFAP. Ganglion cells, neurons, fibroblasts and oligodendrocytes and the tumors derived from these cells will stain negatively. Positive tissue: Astrocytoma/brain.

Immunogen: Bovine spinal cord

Specificity: GFAP

Reactivity: Human, Mouse, Rat

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Not required, Enzyme Digestion: Not required

Expected Staining Pattern: Cytoplasmic
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human, Mouse, Rat
Validated Application: Immunohistochemistry (FFPE)
Host Species: Rabbit
Clonality: Polyclonal
Target Gene: GFAP
Gene ID: 2670
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols