DNA Polymerase I

Catalog number: 18010025

Invitrogen™  Related applications: Restriction Enzyme Cloning

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

DNA Polymerase I is a DNA polymerase with 5´→3´ and 3´→5´ exodeoxyribonuclease activities. DNA Polymerase I also incorporates biotinylated nucleotides.

Applications—DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5´ overhangs
Source—purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Performance and quality testing

Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10 nmol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50 mM potassium phosphate (pH 7.0), 6.7 mM MgCl2 , 1 mM 2-mercaptoethanol, 80 µg/mL template/primer, 32 µM dTTP, 69 nM [3H]dTTP, and enzyme in 100 µL for 30 min at 37°C.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Exonuclease Activity: 3' - 5', 5' - 3'
Hot Start: No
Polymerase: DNA Polymerase I
Product Size: 1,000 units
Shipping Condition: Wet Ice

Contents & storage

1000 units of DNA polymerase in a storage buffer of 50 mM potassium phosphate (pH 7.0), 100 mM KCl, 1 mM DTT, and 50% (v/v) glycerol. It is supplied at 5–10 units/µl.

Store at -5°C to -30°C.

Documents

Manuals & protocols

Recommended products