CD15 Mouse mAb (clone My1), concentrate

Catalog number: 180122

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with human CD15 found on mature granulocytes and on Hodgkin's and Reed-Sternberg cells. The antigen is also present in many carcinomas. CD15 antigen is an epitope involving the carbohydrate sequence 3-fucosyl-N-acetyllactosamine (also termed X hapten). Positive tissue: Spleen.

Clone/PAD: My1

Immunogen: Human Neutrophils

Specificity: CD15

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Not required. Enzyme Digestion: Not Required. Enzyme Digestion (Digest-All™ 1/Ficin) may enhance staining.

Expected Staining Pattern: Cytoplasm and cytoplasmic membrane
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgM, kappa
Clonality: Monoclonal
Target Gene: CD15
Gene ID: 2526
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols

Recommended products