SuperScript® III First-Strand Synthesis System

Catalog number: 18080051

Invitrogen™  Related applications: Reverse Transcription

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The SuperScript® III First-Strand Synthesis System for RT-PCR is optimized to synthesize first-strand cDNA from purified poly(A)+ or total RNA. RNA targets from 100 bp to >12 kb can be detected with this system. The amount of starting material can vary from 1 pg–5 µg of total RNA. SuperScript® III Reverse Transcriptase is a version of M-MLV RT that has been engineered to reduce RNase H activity and provide increased thermal stability. The enzyme is used to synthesize cDNA at a temperature range of 42–55°C, providing increased specificity, higher yields of cDNA, and more full-length product than other reverse transcriptases. Because SuperScript® III RT is not significantly inhibited by ribosomal and transfer RNA, it may be used to synthesize first-strand cDNA from a total RNA preparation.

Using the SuperScript® III First-Strand System
cDNA synthesis is performed in the first step using either total RNA or poly(A)+-selected RNA primed with oligo(dT), random primers, or a gene-specific primer. In the second step, PCR is performed in a separate tube using primers specific for the gene of interest. For the PCR reaction, we recommend one of the following DNA polymerases: Platinum® Taq DNA Polymerase provides automatic hot-start conditions for increased specificity up to 4 kb, Platinum® Taq DNA Polymerase High Fidelity provides increased yield and high fidelity for targets up to 15 kb, and Platinum® Pfx DNA Polymerase provides maximum fidelity for targets up to 12 kb.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reverse Transcriptase: SuperScript® III
Downstream Application: Microarray Analysis, Reverse Transcriptase PCR (RT-PCR)
Reaction Format: Separate Components
Optimal Reaction Temperature: 50° C
Product Line: SuperScript®
Product Size: 50 reactions
Shipping Condition: Dry Ice

Contents & storage

The SuperScript® III First-Strand Synthesis System for RT-PCR is supplied with the following:
• Oligo(dT)20 (50 µM), 50 µl
• Random hexamers (50 ng/µl), 250 µl
• 10X RT buffer, 1 mL
• 0.1 M DTT, 250 µl
• 25 mM magnesium chloride, 500 µl
• 10 mM dNTP mix, 250 µl
• SuperScript III RT (200 U/µl), 50 µl
• RNaseOUT (40 U/µl), 100 µl
E. coli RNase H (2 U/µl), 50 µl
• DEPC-treated water, 1.2 mL
• Total HeLa RNA (10 ng/µl), 20 µl
• Sense Control Primer (10 µM), 25 µl
• Antisense Control Primer (10 µM), 25 µl

Sufficient for 50 reactions, based on a 20 µl reaction size. Store at -20°C.

Documents

Manuals & protocols

Recommended products