CD10 Mouse mAb (clone 56C6), concentrate

Catalog number: 182287

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with human CD10. CD10 antigen is expressed in a wide variety of epithelial and non epithelial tissues, but its most significant application is in the diagnosis and classification of certain types of malignant lymphoma and leukemia. CD10 is expressed in a high percentage of cases of acute lymphoblastic leukemia (ALL), follicular lymphoma, Burkitt's lymphoma, and some hematopoietic tumors.Positive tissue: Normal kidney/smallintestine.

Clone/PAD: 56C6

Immunogen: Prokaryotic recombinant fusion protein corresponding to the external domain of the CD10 glycoprotein.

Specificity: CD10

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (EDTA Buffer pH 8.0), Enzyme Digestion: Not required

Expected Staining Pattern: Membrane
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgG1
Clonality: Monoclonal
Target Gene: CD10
Gene ID: 4311
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 0.2 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols