Lipofectin™ Transfection Reagent

Catalog number: 18292037

Invitrogen™  Related applications: Transfection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Lipofectin® Transfection Reagent is the reagent of choice for transfection of endothelial cells. Lipofectin® Transfection Reagent is also suitable for transfecting DNA, RNA, and oligonucleotides into mammalian cells, and DNA into plant protoplasts. Lipofectin® reagent has also been shown to work well, in combination with PLUS® Reagent, for the transfection of HeLa cells. Lipofectin® Transfection Reagent is a 1:1 (w/w) liposome formulation of the cationic lipid N-[1-(2,3-dioleyloxy)propyl]-n,n,n-trimethylammonium chloride (DOTMA) and dioleoyl phophotidylethanolamine (DOPE) in membrane-filtered water.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Established Cell Lines, Hard-to-Transfect Cells, Primary Cells
Sample Type (Specific): Plasmid DNA
Transfection Technique: Lipid-Based Transfection
Serum Compatible: No
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Product Line: Lipofectin™
Product Size: 4 mL
Shipping Condition: Wet Ice

Contents & storage

Each unit contains one or multiple vials (1 ml) of reagent. Store at +4°C. Lipofectin® Transfection Reagent is guaranteed stable for 6 months when stored properly.

Documents

Manuals & protocols

Recommended products