MAX Efficiency™ DH10B™ Competent Cells

Catalog number:  18297010

Invitrogen™  Related applications: Transformation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

MAX Efficiency® DH10B™ Competent Cells are highly efficient E. coli, perfect for most applications. DH10B™ E. coli are available in both electrocompetent and chemically competent formats. The high transformation efficiency makes them ideal for generating cDNA or genomic libraries. High-efficiency transformation is seen with plasmids 150 kb in size. The versatile DH10B™ genotype has the following features:

lacZ for blue/white screening of recombinant clones
• Elimination of mcrA, mcrBC, mrr, and hsdRMS restriction systems to allow construction of more representative genomic libraries
• A1 mutation to increase plasmid yield and quantity
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Competent Cell Type: Chemically Competent
Species: E. coli
Format: Tube(s)
Transformation Efficiency: > 1x10^9
Bacterial or Yeast Strain: DH10B™
Product Line: DH10B™, MAX Efficiency®
Blue-White Screening: Yes
Cloning Methylated DNA: Yes
Cloning Unstable DNA: Not Suitable for cloning unstable DNA
F' Episome (to make ssDNA): Lacks F' Episome
High Throughput Compatibility: Not High Throughput-Compatible (Manual)
Improves Plasmid Quality: Yes
Preparing Unmethylated DNA: Not Suitable for preparing unmethylated DNA
Product Size: 10 reactions
Propagating ccdB Vectors: Not for ccdB vector propagation
Reduces Recombination: Yes
T1 Phage - Resistant (tonA): No
Shipping Condition: Dry Ice

Contents & storage

Contains:
• MAX Efficiency® DH10B™ Competent Cells: 5 vials, 200 µl each (total of 1 ml)
• pUC19 DNA (0.01 µg/ml): 1 vial, 100 µl
• S.O.C. medium: 2 bottles, 6 ml each

Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature.

Documents

Manuals & protocols

Recommended products